missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GDF-3 Monoclonal antibody specifically detects GDF-3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | GDF-3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SR1219 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human GDF-3 (Uniprot #: Q9NR23) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?