missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-3 Antibody (010), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89220B
This item is not returnable.
View return policy
Description
GDF-3 Monoclonal specifically detects GDF-3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GDF-3 | |
| Monoclonal | |
| Biotin | |
| GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human GDF-3. | |
| 0.1 mL | |
| Cytokine Research, Stem Cell Markers | |
| 9573 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 10 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction