missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GDF-15 Antibody (ponsegromab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | GDF-15 |
|---|---|
| Clone | ponsegromab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047646
|
Novus Biologicals™
NBP3-28880-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045323
|
Novus Biologicals™
NBP3-28880-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044598
|
Novus Biologicals™
NBP3-28880-100ug |
100 μg |
£266.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-15 Monoclonal antibody specifically detects GDF-15 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| GDF-15 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Hepatic Stem Cell Markers, Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
| GDF15/ MIC1 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| ponsegromab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q99988 | |
| 9518 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title