missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
£190.00 - £459.00
Specifications
| Antigen | GDF-15 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation Reactivity reported in (PMID: 26114631)., Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18447830
|
Novus Biologicals
NBP1-81050-25ul |
25ul |
£190.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18751534
|
Novus Biologicals
NBP1-81050 |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
GDF-15 Polyclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GDF-15 | |
| Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
| GDF15 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation Reactivity reported in (PMID: 26114631)., Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q99988 | |
| 9518 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSLAEASRASFPGPSELHSEDSRFRELRKRYEDLLTRLRANQSWEDSNTDLVPAPAVRILTPEVRLGSGGHLHLRISRAALPEGLPEASRLHRALFRLSPTASRSWDVTRPLRRQLSLARPQAPALHLRLSPPPSQSDQL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 34 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title