missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GDF-11/GDF-8 Antibody (apitegromab) - IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Especificaciones

Antigen GDF-11/GDF-8
Clone apitegromab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
Ver más especificaciones

Productos 3
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
30048146
Ver documentos
Novus Biologicals™
NBP3-28425-1mg
1 mg
£695.00
1mg
Ver productos alternativos
Ahorre hasta 
»
Please to purchase this item. Need a web account? Register with us today!
30046507
Ver documentos
Novus Biologicals™
NBP3-28425-50ug
50 μg
£162.00
50µg
Ver productos alternativos
Ahorre hasta 
»
Please to purchase this item. Need a web account? Register with us today!
30045599
Ver documentos
Novus Biologicals™
NBP3-28425-100ug
100 μg
£247.00
100µg
Ver productos alternativos
Ahorre hasta 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GDF-11/GDF-8 Monoclonal antibody specifically detects GDF-11/GDF-8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

GDF-11/GDF-8
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Human
O14793
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
apitegromab
Monoclonal
Purified
RUO
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
GDF8/ Myostatin
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.