missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GDF-11/BMP-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £391.00
Specifications
| Antigen | GDF-11/BMP-11 |
|---|---|
| Clone | 937320 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797139
![]() |
R&D Systems
MAB19583-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18798089
![]() |
R&D Systems
MAB19583 |
100 μg |
£391.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-11/BMP-11 Monoclonal specifically detects GDF-11/BMP-11 in Human samples. It is validated for Immunocytochemistry.Specifications
| GDF-11/BMP-11 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11 | |
| GDF11 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 937320 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95390 | |
| 10220 | |
| E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GDF-11/BMP-11 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title