missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF-11/BMP-11 Antibody (SR1761), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | GDF-11/BMP-11 |
|---|---|
| Clone | SR1761 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680685
|
Novus Biologicals
NBP3-21706-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650874
|
Novus Biologicals
NBP3-21706-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-11/BMP-11 Monoclonal antibody specifically detects GDF-11/BMP-11 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| GDF-11/BMP-11 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| BMP-11BMP11Bone morphogenetic protein 11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11 | |
| A synthesized peptide derived from human GDF-11/BMP-11 (Uniprot #: O95390) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1761 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10220 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title