missing translation for 'onlineSavingsMsg'
Learn More

GDF-1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18372491 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372491 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372491 Supplier Novus Biologicals Supplier No. NBP281826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GDF-1 Polyclonal specifically detects GDF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF-1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias DORV, DTGA3, embryonic growth/differentiation factor 1, GDF-1, growth differentiation factor 1
Gene Symbols GDF1
Host Species Rabbit
Immunogen Rabbit polyclonal GDF-1 antibody was raised against a 16 amino acid peptide near the amino terminus of human GDF-1. The immunogen is located within amino acids 30 - 80 of GDF-1. Amino Acid Squence: CDEPQGAPRLRPVPPVM
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2657
Test Specificity GDF-1 antibody is predicted to not cross-react with any other members of the growth differentiation factor family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.