missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11272-2-AP-150UL
This item is not returnable.
View return policy
Description
Has glycerophosphoinositol phosphodiesterase activity. Has little or no activity towards glycerophosphocholine. GDE1 activity can be modulated by G-protein signaling pathways.Specifications
| GDE1 | |
| Polyclonal | |
| Unconjugated | |
| GDE1 | |
| 363E6.2, GDE1, MIR16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51573, 56209, 60418 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JL55, Q9JL56, Q9NZC3 | |
| GDE1 | |
| GDE1 Fusion Protein Ag1821 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction