missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDC-0980, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | (S)-1-[4-[(2-(2-Aminopyrimidin-5-yl)-7-methyl-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-6-yl)methyl]piperazin-1-yl]-2-hydroxypropan-1-one |
| CAS | 1032754-93-0 |
| Molecular Formula | C23H30N8O3S |
| Quantity | 50 mg |
| Synonym | GDC-0980, Apitolisib, G 038390, G 038390.1, GDC 0980, GDC 0980.1, GNE 390, RG 7422 |
| InChI Formula | InChI=1S/C23H30N8O3S/c1-14-17(13-29-3-5-31(6-4-29)22(33)15(2)32)35-19-18(14)27-20(16-11-25-23(24)26-12-16)28-21(19)30-7-9-34-10-8-30/h11-12,15,32H,3-10,13H2,1-2H3,(H2,24,25,26)/t15-/m0/s1 |
| SMILES | C[C@H](O)C(=O)N1CCN(Cc2sc3c(nc(nc3c2C)c4cnc(N)nc4)N5CCOCC5)CC1 |
| IUPAC Name | (2S)-1-[4-[[2-(2-aminopyrimidin-5-yl)-7-methyl-4-morpholin-4-ylthieno[3,2-d]pyrimidin-6-yl]methyl]piperazin-1-yl]-2-hydroxypropan-1-one |
| Molecular Weight (g/mol) | 498.6 |
| Formula Weight | 498.2162 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction