missing translation for 'onlineSavingsMsg'
Learn More

GDC-0927, MedChemExpress

Product Code. 16493096
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493096 25 mg 25mg
16487187 5 mg 5mg
16480777 50 mg 50mg
16444468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493096 Supplier Medchem Express Supplier No. HY111484
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GDC-0927 (SRN-927) is a potent, non-steroidal, orally bioavailable, selective estrogen receptor antagonist.

GDC-0927 (SRN-927) is a potent, non-steroidal, orally bioavailable, selective estrogen receptor antagonist.

Specifications

Chemical Name or Material GDC-0927
CAS 1642297-01-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C28H28FNO4
Quantity 25 mg
Synonym SRN-927
Solubility Information DMSO : 83.33 mg/mL (180.56 mM; Need ultrasonic)
SMILES OC1=CC=C2C(C(C)=C(C3=CC=CC(O)=C3)[C@H](C4=CC=C(OCCN5CC(CF)C5)C=C4)O2)=C1
Molecular Weight (g/mol) 461.52
Formula Weight 461.52
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.