missing translation for 'onlineSavingsMsg'
Learn More

GDC-0879, MedChemExpress

Product Code. 16473756
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473756 25 mg 25mg
16460578 5 mg 5mg
16458118 10 mg 10mg
16434037 50 mg 50mg
16420356 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473756 Supplier Medchem Express Supplier No. HY50864
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GDC-0879 is a potent and selective B-Raf inhibitor with an IC50 of 0.13 nM.

GDC-0879 is a potent and selective B-Raf inhibitor with an IC50 of 0.13 nM.

Specifications

Chemical Name or Material GDC-0879
CAS 905281-76-7
Color Light Nude
Purity Grade Notes Research
Molecular Formula C19H18N4O2
Quantity 25 mg
Solubility Information DMSO : 50 mg/mL (149.53 mM; Need ultrasonic)
SMILES O/N=C1C(C=CC(C2=CN(CCO)N=C2C3=CC=NC=C3)=C4)=C4CC/1
Molecular Weight (g/mol) 334.37
Formula Weight 334.37
Percent Purity 99.07%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.