missing translation for 'onlineSavingsMsg'
Learn More

GDC-0879 Hydrochloride, TRC

Product Code. 30396381
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
250 mg
Unit Size:
2.5mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396381 25 mg 25mg
30399084 250 mg 250mg
30399664 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396381 Supplier Toronto Research Chemicals Supplier No. TRCG29998025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydro-5-[1-(2-hydroxyethyl)-3-(4-pyridinyl)-1H-pyrazol-4-yl]-1H-Inden-1-one Oxime Hydrochloride
Analyte or Component Names 2,3-Dihydro-5-[1-(2-hydroxyethyl)-3-(4-pyridinyl)-1H-pyrazol-4-yl]-1H-Inden-1-one Oxime Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19H19ClN4O2
Synonym GDC-0879 Hydrochloride
InChI Formula InChI=1S/C19H18N4O2/c24-10-9-23-12-17(19(21-23)13-5-7-20-8-6-13)15-1-3-16-14(11-15)2-4-18(16)22-25/h1,3,5-8,11-12,24-25H,2,4,9-10H2/b22-18-
Solution Type Neat
SMILES OCCn1cc(c2ccc3\C(=N\O)\CCc3c2)c(n1)c4ccncc4
Industry Type Pharmaceutical
Molecular Weight (g/mol) 370.83
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.