missing translation for 'onlineSavingsMsg'
Learn More

GDC-0834 S-enantiomer, MedChemExpress

Product Code. 16439097
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439097 5 mg 5mg
16406518 10 mg 10mg
16400246 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16439097 Supplier Medchem Express Supplier No. HY15427B
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GDC-0834 (S-enantiomer) is the S-enantiomer of GDC-0834. GDC-0834 is a potent and selective BTK inhibitor.

GDC-0834 (S-enantiomer) is the S-enantiomer of GDC-0834. GDC-0834 is a potent and selective BTK inhibitor.

Specifications

Chemical Name or Material GDC-0834 S-enantiomer
Physical Form Solid
CAS 1133432-50-4
Color Reddish Brown
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C33H36N6O3S
Solubility Information DMSO : 100 mg/mL (167.58 mM; Need ultrasonic)
SMILES CC1=C(C(N=C2NC3=CC=C([C@@H]4N(C)CCN(C)C4=O)C=C3)=CN(C)C2=O)C=CC=C1NC(C5=CC6=C(CCCC6)S5)=O
Molecular Weight (g/mol) 596.74
Formula Weight 596.74
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.