missing translation for 'onlineSavingsMsg'
Learn More

GDC-0834, MedChemExpress

Product Code. 16486508
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486508

Brand: Medchem Express HY15427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0834 is a potent and selective BTK inhibitor. GDC-0834 inhibits BTK with an in vitro IC50 of 5.9 and 6.4 nM in biochemical and cellular assays, respectively, and in vivo IC50 of 1.1 and 5.6 μM in mouse and rat, respectively.

GDC-0834 is a potent and selective BTK inhibitor. GDC-0834 inhibits BTK with an in vitro IC50 of 5.9 and 6.4 nM in biochemical and cellular assays, respectively, and in vivo IC50 of 1.1 and 5.6 μM in mouse and rat, respectively.

Specifications

Chemical Name or Material GDC-0834
Physical Form Solid
CAS 1133432-49-1
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C33H36N6O3S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 32 mg/mL (53.62 mM)
SMILES O=C(C1=CC(CCCC2)=C2S1)NC3=CC=CC(C(N=C4NC5=CC=C([C@H]6N(C)CCN(C)C6=O)C=C5)=CN(C)C4=O)=C3C
Molecular Weight (g/mol) 596.74
Formula Weight 596.74
Percent Purity 98.21%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.