missing translation for 'onlineSavingsMsg'
Learn More

GDC-0623, MedChemExpress

Product Code. 16499237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499237

Brand: Medchem Express HY15610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GDC-0623 (RG 7421) is a potent, ATP-uncompetitive inhibitor of MEK1 (Ki=0.13 nM, +ATP), and displays 6-fold weaker potency against HCT116 (KRAS (G13D), EC50=42 nM) versus A375 (BRAFV600E, EC50=7 nM).

GDC-0623 (RG 7421) is a potent, ATP-uncompetitive inhibitor of MEK1 (Ki=0.13 nM, +ATP), and displays 6-fold weaker potency against HCT116 (KRAS (G13D), EC50=42 nM) versus A375 (BRAFV600E, EC50=7 nM).

Specifications

Chemical Name or Material GDC-0623
CAS 1168091-68-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H14FIN4O3
Quantity 5 mg
Synonym RG 7421 MEK inhibitor 1
Solubility Information DMSO : 50 mg/mL (109.60 mM; Need ultrasonic)
SMILES FC(C=C(I)C=C1)=C1NC2=C(C(NOCCO)=O)C=CC3=CN=CN23
Molecular Weight (g/mol) 456.21
Formula Weight 456.21
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.