missing translation for 'onlineSavingsMsg'
Learn More

GDC-0349, MedChemExpress

Product Code. 16436398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16436398 10 mg 10mg
16428987 5 mg 5mg
16422507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16436398 Supplier Medchem Express Supplier No. HY15248
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GDC-0349 is a potent and selective ATP-competitive mTOR inhibitor with a Ki of 3.8 nM. GDC-0349 inhibits of both mTORC1 and mTORC2 complexes.

GDC-0349 is a potent and selective ATP-competitive mTOR inhibitor with a Ki of 3.8 nM. GDC-0349 inhibits of both mTORC1 and mTORC2 complexes.

Specifications

Chemical Name or Material GDC-0349
CAS 1207360-89-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H32N6O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (220.97 mM)
SMILES O=C(NCC)NC(C=C1)=CC=C1C2=NC3=C(CCN(C4COC4)C3)C(N5[C@@H](C)COCC5)=N2
Molecular Weight (g/mol) 452.55
Formula Weight 452.55
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.