missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561175
This item is not returnable.
View return policy
Description
Immunogen sequence: LNSSDTTAEI FQEDTVRSPF LYNKDVNGKV VLWKGDVALL NCTAIVNTSN ESLTDKNPVS ESIFMLAGPD LKEDLQKLKG CRTGEAKL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.
GDAP2 is a protein coding gene and gene ontology (GO) annotation includes lysosomal membrane; response to retinoic acid.
Specifications
| GDAP2 | |
| Polyclonal | |
| Unconjugated | |
| GDAP2 | |
| C77050; D3Ertd801e; dJ776P7.1; ganglioside induced differentiation associated protein 2; ganglioside-induced differentiation-associated protein 2; Ganglioside-induced differentiation-associated-protein 2; GDAP2; MACROD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NXN4 | |
| GDAP2 | |
| Recombinant protein corresponding to Human GDAP2. Recombinant protein control fragment (Product #RP-94543). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction