missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDAP1 Mouse anti-Human, Mouse, Rat, Rabbit, Pig, Clone: 3E10D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68083-1-IG-20UL
This item is not returnable.
View return policy
Description
GDAP1 Monoclonal antibody apecifically detetects GDAP1 antigen in Human, Mouse, Rat, Rabbit, Pig Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| GDAP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| GDAP1 | |
| GDAP1 fusion protein Ag28616 | |
| 20 μL | |
| 54332 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG2b |
| Western Blot, Immunohistochemistry, ELISA | |
| 3E10D4 | |
| Unconjugated | |
| BC024939 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction