missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119167
This item is not returnable.
View return policy
Description
GDA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications
| GDA | |
| Polyclonal | |
| Unconjugated | |
| GDA | |
| AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 14544, 525937, 9615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9R111, Q9Y2T3 | |
| GDA | |
| Recombinant protein Guanine Deaminase. The antigen corresponds to amino acid range 1-454 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction