Learn More
Invitrogen™ GDA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592978
Description
Immunogen sequence: FVGKVCMDLN DTFPEYKETT EESIKETERF VSEMLQKNYS RVKPIVTPRF SLSCSETLMG ELGNIAKTRD LHIQSHISEN RDEVEAVKNL YPSYKNYTSV YDKNNLLTNK TVMAHGCYLS AEELNVFHER GASIAHCPNS NLSLSSGFLN VLEVLKHEVK IGLGTDVAGG YSYSMLDAIR RAVMVSNILL INKVNEKSLT LKEVFRLATL GGSQALGLDG EIGNFEVGKE FDAILINPKA SDSPIDLFYG DFFGDISEAV IQKFLYLGDD RNIEEVYVGG KQVVPFSSSV; Positive Samples: HeLa, SKOV3, mouse liver, mouse brain, rat brain, Mouse lung, Rat testis, NCI-H460.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications
| GDA | |
| Polyclonal | |
| Unconjugated | |
| GDA | |
| AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14544, 83585, 9615 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R111, Q9WTT6, Q9Y2T3 | |
| GDA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 165-454 of human GDA (NP_0042841). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.