missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554976
This item is not returnable.
View return policy
Description
Immunogen sequence: NKVNEKSLTL KEVFRLATLG GSQALGLDGE IGNFEVGKEF DAILINPKAS DSPIDLFYGD FFGDISEAVI QKFLYLGDDR NIEEVYVGGK QVVPF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications
| GDA | |
| Polyclonal | |
| Unconjugated | |
| GDA | |
| AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2T3 | |
| GDA | |
| Recombinant protein corresponding to Human GDA. Recombinant protein control fragment (Product #RP-92913). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction