missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCSH Recombinant Rabbit Monoclonal Antibody (5U2H0)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | GCSH |
|---|---|
| Clone | 5U2H0 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283389
|
Invitrogen™
MA558155 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283725
![]() |
Invitrogen™
MA558156 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: SVRKFTEKHE WVTTENGIGT VGISNFAQEA LGDVVYCSLP EVGTKLNKQD EFGALESVKA ASELYSPLSG EVTEINEALA ENPGLVNKSC YEDGWLIKMT LSNPSELDEL MSEEAYEKYI KSIEE.
The glycine cleavage system catalyzes the degradation of glycine. The H protein (GCSH) shuttles the methylamine group of glycine from the P protein (GLDC) to the T protein (GCST).Specifications
| GCSH | |
| 1 mg/mL | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P23434 | |
| 2653 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 49-173 of human GCSH (NP_004474.2). | |
| Antibody |
| 5U2H0 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| GCSH | |
| 1100001L02Rik; 5730591C18Rik; GCE; Gcsh; Glycine cleavage system H protein, mitochondrial; glycine cleavage system protein H; glycine cleavage system protein H (aminomethyl carrier); lipoic acid-containing protein; mitochondrial glycine cleavage system H-protein; NKH | |
| GCSH | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title