missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCSAML Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555639
This item is not returnable.
View return policy
Description
Immunogen sequence: MGNYLLRKLS CLGENQKKPK KGNPDEERKR QEMTTFERKL QDQDKKSQEV SSTSNQENEN GSGSEEV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.
This gene encodes a protein thought to be a signaling molecule associated with germinal centers, the sites of proliferation and differentiation of mature B lymphocytes. Alternate splicing results in multiple transcript variants.
Specifications
| GCSAML | |
| Polyclonal | |
| Unconjugated | |
| GCSAML | |
| C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 148823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JQS6 | |
| GCSAML | |
| Recombinant protein corresponding to Human GCSAML. Recombinant protein control fragment (Product #RP-94447). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction