missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCSAM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544375
This item is not returnable.
View return policy
Description
Peptide sequence: YSLLHMPSTD PRHARSPEDE YELLMPHRIS SHFLQQPRPL MAPSETQFSH Sequence homology: Human: 100%.
GCET2 is a protein which may function in signal transduction pathways and whose expression is elevated in germinal cell lymphomas. It contains a putative PDZ-interacting domain, an immunoreceptor tyrosine-based activation motif (ITAM), and two putative SH2 binding sites. In B cells, its expression is specifically induced by interleukin-4.This gene encodes a protein which may function in signal transduction pathways and whose expression is elevated in germinal cell lymphomas. It contains a putative PDZ-interacting domain, an immunoreceptor tyrosine-based activation motif (ITAM), and two putative SH2 binding sites. In B cells, its expression is specifically induced by interleukin-4. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| GCSAM | |
| Polyclonal | |
| Unconjugated | |
| GCSAM | |
| GAL; GCAT2; Gcet; GCET2; GCET2 germinal center expressed transcript 2; GCSAM; Germinal center associated lymphoma (hGAL); germinal center associated signaling and motility; germinal center associated, signaling and motility; germinal center B-cell-expressed transcript 2 protein; germinal center expressed transcript 2; germinal center-associated lymphoma protein; Germinal center-associated signaling and motility protein; germinal center-associated, signaling and motility; HGAL; human germinal center-associated lymphoma; M17; M17-L; MGC40441 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 257144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N6F7 | |
| GCSAM | |
| Synthetic peptide directed towards the middle region of human GCET2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction