missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GCS1 Polyclonal specifically detects GCS1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | GCS1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | GCS1, glucosidase I, mannosyl-oligosaccharide glucosidase, processing A-glucosidase I |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human GCS1 (NP_006293.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?