missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCS1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588726

Product Code. 16546861

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RWVLAWYRAR RAVTLHSAPP VLPADSSSPA VAPDLFWGTY RPHVYFGMKT RSPKPLLTGL MWAQQGTTPG TPKLRHTCEQ GDGVGPYGWE FHDGLSFGRQ HIQDGALRLT TEFVKRPGGQ HGGDWSWRVT VEPQDSGTSA LPLVSLFFYV VTDGKEVLLP EVGAKGQLKF ISGHTSELGD FRFTLLPPTS PGDTAPKYGS YNVFWTSNPG LPLLTEMVKS RLNSWFQHRP PGAPPERYLG LPGSLKWEDR GPSGQGQGQF L; Positive Samples: U-87MG, LO2, BxPC-3, OVCAR-3, Mouse Brain, Mouse Liver, Mouse Pancreas, Mouse Testis, Rat Liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.

GCS1 cleaves the distal alpha 1,2-linked glucose residue from the Glc(3)Man(9)GlcNAc(2) oligosaccharide precursor in a highly specific manner. Defects in GCS1 are the cause of type IIb congenital disorder of glycosylation (CDGIIb). This syndrome is also known as glucosidase I deficiency and is characterized by marked generalized hypotonia and hypomotility of the neonate, dysmorphic features, including a prominent occiput, short palpebral fissures, retrognathia, high arched palate, generalized edema, and hypoplastic genitalia. Symptoms include hepatomegaly, hypoventilation, feeding problems and seizures. The clinical course is progressive and survival is at most a few months.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCS1
Polyclonal
Unconjugated
MOGS
1810017N02Rik; AI181835; CDG2B; CWH41; DER7; GCS1; Glucosidase 1; glucosidase I; glycoprotein-processing glucosidase I; Mannosyl-oligosaccharide glucosidase; Mogs; Processing A-glucosidase I
Rabbit
Affinity Chromatography
RUO
57377, 7841
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.70 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q13724, Q80UM7
MOGS
Recombinant fusion protein containing a sequence corresponding to amino acids 60-320 of human MOGS (NP_0062932).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.