missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14967
This item is not returnable.
View return policy
Description
GCS1 Polyclonal specifically detects GCS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.
Specifications
| GCS1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| GCS1, glucosidase I, mannosyl-oligosaccharide glucosidase, processing A-glucosidase I | |
| Rabbit | |
| 92 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence Reported in scientific literature (PMID:29465311) , Immunoprecipitation Validated from a verified customer review., Knockdown Validated Reported in scientific literature (PMID:29465311) | |
| Q13724 | |
| MOGS | |
| Recombinant protein encompassing a sequence within the center region of human Glucosidase I. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 7841 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction