missing translation for 'onlineSavingsMsg'
Learn More

GCR2 Mouse anti-Human, Clone: PCRP-ECD-1D10, Novus Biologicals™

Product Code. 18365014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365014 100 μg 100µg
18351684 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18365014 Supplier Novus Biologicals Supplier No. NBP313849100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GCR2 Monoclonal antibody specifically detects GCR2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCR2
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-ECD-1D10
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. O95905
Gene Alias ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X
Host Species Mouse
Immunogen Recombinant full-length human GCR2 protein (Uniprot: O95905)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11319
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.