missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GCR2 Polyclonal specifically detects GCR2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | GCR2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse GCR2. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?