Learn More
Invitrogen™ GCP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116646
Description
Positive Samples: A-549, Mouse brain Immunogen sequence: KTINLLKLCC PRHYLCWSDV PVPRISVIFS LEELKEIEKD CAVYVGRMER VARHSSVSKE EKELRMEIAK QELIAHAREA ASRVLSALSD RQMSERMALD ARKREQFQRL KEQFVKDQER RQAARQEELD DDFSYARELR DRERRLKSLE EELERKARQA LVDHYSKLSA EAARREQKAL WRIQRHRLES ARLRFLLEDE KHIQEMLKAV SEAHQPQEPP DVLLSVHPQV TSPGPEHPEG GQGCDSGSAE QHSPAWDGWN RPGLLTPQPL KPLAVGAGGR GLQQAEGARP FSDSLSIGDF L.
Tubulin Gamma Complex Associated Protein 6 (GCP6) is part of a large multisubunit complex required for microtubule nucleation at the centrosome.
Specifications
| GCP6 | |
| Polyclonal | |
| Unconjugated | |
| TUBGCP6 | |
| AI843275; D430023H11; Gamma-tubulin complex component 6; GCP6; GCP-6; KIAA1669; MCCRP; MCCRP1; MCPHCR; mKIAA1669; TUBGCP6; tubulin gamma complex associated protein 6; tubulin, gamma complex associated protein 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 328580, 362980, 85378 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.79 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| G5E8P0, Q96RT7 | |
| TUBGCP6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-900 of human TUBGCP6 (NP_0651942). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.