missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCP4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA575415

Product Code. 15911945

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complexassociated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP4 (gamma-Tubulin complex component 4), also known as TUBGCP4, is a ubiquitously expressed 667 amino acid member of the gamma-Tubulin complex that localizes to the metaphase spindle during mitosis. In response to proteosome inhibition, GCP4 exhibits increased accumulation at the pericentiolar material where it participates in microtubule organization and nucleation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCP4
Polyclonal
Unconjugated
TUBGCP4
4932441P04Rik; 76P; D2Ertd435e; gamma tubulin ring complex protein; gamma tubulin ring complex protein (76p gene); gamma-ring complex protein 76 kDa; Gamma-tubulin complex component 4; GCP4; GCP-4; Grip76; h76p; hGCP4; hGrip76; MCCRP3; RGD1306924; TUBGCP4; tubulin gamma complex associated protein 4; tubulin, gamma complex associated protein 4
Rabbit
Antigen Affinity Chromatography
RUO
27229, 362203, 51885
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Q9D4F8, Q9UGJ1
TUBGCP4
Synthetic peptide corresponding to amino acids 197-243 of Human GCP4.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.