missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCNT4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103649
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GCNT4.
Glycosyltransferase that mediates core 2 O-glycan branching, an important step in mucin-type biosynthesis. Does not have core 4 O-glycan or I-branching enzyme activity.
Specifications
| GCNT4 | |
| Polyclonal | |
| Unconjugated | |
| GCNT4 | |
| 6-N-acetylg; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 4; C2GNT3; core 2 beta-1,6-N-acetylglucosaminyltransferase 3; core 2-branching enzyme 3; Core2-GlcNAc-transferase 3; Gcnt4; glucosaminyl (N-acetyl) transferase 3, mucin type pseudogene; glucosaminyl (N-acetyl) transferase 4; glucosaminyl (N-acetyl) transferase 4, core 2; glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase); glucosaminyl transferase 4, core 2; Gm279; Gm73 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 218476, 51301 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9Q649, Q9P109 | |
| GCNT4 | |
| A synthesized peptide derived from human GCNT4(Accession Q9P109), corresponding to amino acid residues D100-I150. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction