missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCNT3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | GCNT3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18337394
|
Novus Biologicals
NBP3-04915-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18395584
|
Novus Biologicals
NBP3-04915-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCNT3 Polyclonal specifically detects GCNT3 in Human samples. It is validated for Western Blot.Specifications
| GCNT3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N-acetylglucosaminyltransferase 3, beta1,6GlcNAc-transferase, beta1,6-N-acetylglucosaminyltransferase-M, C2/4GnTGNTM, C24GNT, C2GnT2, C2GnT-M, C2GNTM, C2GnT-mucin type, Core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase, EC 2.4.1.102, EC 2.4.1.150, glucosaminyl (N-acetyl) transferase 3, mucin type, hC2GnT-M, mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-180 of human GCNT3 (NP_004742.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mucosal Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 9245 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title