missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCNT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18118-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the enzyme responsible for formation of the blood group I antigen. The i and I antigens are distinguished by linear and branched poly-N-acetyllactosaminoglycans, respectively. The encoded protein is the I-branching enzyme, a beta-1, 6-N-acetylglucosaminyltransferase responsible for the conversion of fetal i antigen to adult I antigen in erythrocytes during embryonic development. Mutations in this gene have been associated with adult i blood group phenotype. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| GCNT2 | |
| Polyclonal | |
| Unconjugated | |
| GCNT2 | |
| CCAT, GCNT2, GCNT2C, GCNT5, I branching enzyme, IGNT, II, NACGT1, NAGCT1, ULG3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14538, 2651, 306860 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97402, Q8N0V5 | |
| Gcnt2 | |
| GCNT2 Fusion Protein Ag12635 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction