missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCN5 Monoclonal Antibody (2GC-2C11)
GREENER_CHOICE

Product Code. 15652022
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15652022 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15652022 Supplier Invitrogen™ Supplier No. MA3046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA3-046 detects GCN5 in human samples. MA3-046 has been succesfully used in western blot and ChIP applications. Immunogen is a synthetic peptide corresponding to residues 118-132 of human GCN5.

SAGA (Spt-Ada-Gen5 histone acetyltransferase complex) is a histoneacetylase complex which has Gcn5p as catalyst subunit and functions overlapping with the fundamental transcription factor TFIID which has Taf1p as catalyst subunit. However, SAGA and TFIID have different allotment and each accomplishes the important role in the transcription for Housekeeping gene group and Stress Responding gene group. Also, SAGA is a gigantic protein complex which is composed of Ada protein group (5 kinds), TBP related protein group (4 kinds), TAF protein group (5 kinds that also pertain to TFIID), and other protein groups (>6 kinds). As its molecular function, there is the chemical modification of histone, or the recruitment by direct interaction of transcription regulating factor on DNA, or the control of transcription starting reaction by TBP. Gcn5p that show histoneacetylase activity is one kind of the above-mentioned Ada protein group, and in the case of budding yeast it is composed of 439 amino acid residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCN5
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone 2GC-2C11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene KAT2A
Gene Accession No. Q92830
Gene Alias 1110051E14Rik; AAS104; ADA4; AW212720; GCN5; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2; GCN5 general control of amino acid synthesis-like 2; Gcn5l2; general control of amino acid synthesis 5-like 2; general control of amino acid synthesis protein 5-like 2; general control of amino acid synthesis, yeast homolog-like 2; General control of amino acid synthesis, yeast, homolog-like 2; general control of amino acid synthesis-like 2; hGCN5; Histone acetyltransferase GCN5; histone acetyltransferase KAT2A; Histone succinyltransferase KAT2A; hsGCN5; K(lysine) acetyltransferase 2A; KAT2; Kat2a; KATA2; Lysine acetyltransferase 2A; MmGCN5; PCAF-b; PCAF-B/GCN5; STAF97; SWI9; YGR252W
Gene Symbols KAT2A
Host Species Mouse
Immunogen Synthetic peptide coupled to ovalbumin corresponding to amino acids 118-132 of human GCN5: CKCNGWKNPKPPTAP.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.