missing translation for 'onlineSavingsMsg'
Learn More

GCN4 Antibody (C11L34), DyLight 488, Novus Biologicals™

Product Code. 18371931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371931 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371931 Supplier Novus Biologicals Supplier No. NBP281273G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GCN4 Monoclonal specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCN4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone C11L34
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4
Gene Symbols GCN4
Host Species Mouse
Immunogen Biotinylated GCN4-peptide (HLENEVARLKK).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856709
Test Specificity This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity.
Target Species Yeast
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.