missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN2iB, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
GCN2iB is an ATP-competitive inhibitor of a serine/threonine-protein kinase general control nonderepressible 2 (GCN2), with an IC50 of 2.4 nM.
Specifications
Specifications
| Chemical Name or Material | GCN2iB |
| CAS | 2183470-12-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H12ClF2N5O3S |
| Quantity | 25 mg |
| Solubility Information | DMSO : 16.67 mg/mL (36.89 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC(Cl)=CN=C1OC)(NC2=CC=C(F)C(C#CC3=CN=C(N)N=C3)=C2F)=O |
| Molecular Weight (g/mol) | 451.83 |
| Formula Weight | 451.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction