missing translation for 'onlineSavingsMsg'
Learn More

GCN2 [p Thr899] Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30730995 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30730995 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730995 Supplier Novus Biologicals Supplier No. AF7605MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GCN2 Polyclonal antibody specifically detects GCN2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCN2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1
Host Species Rabbit
Immunogen Phosphopeptide containing human GCN2 T899 site
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 440275
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.