missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1593B
This item is not returnable.
View return policy
Description
GCN2 Polyclonal antibody specifically detects GCN2 in Human samples. It is validated for Western Blot
Specifications
| GCN2 | |
| Polyclonal | |
| Western Blot | |
| eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human GCN2 (eIF2AK4, eukaryotic translation initiation factor 2 alpha kinase 4) using the numbering given in entry NP_001013725.2 (GeneID 440275). | |
| 0.1 mL | |
| Phospho Specific, Protein Kinase | |
| 440275 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction