missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCN1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553848
This item is not returnable.
View return policy
Description
Immunogen sequence: EALVTDAGEV TEAGKAYVPP RVLQEALCVI SGVPGLKGDV TDTEQLAQEM LIISHHPSLV AVQSGLWPAL LARMKIDPEA FITRHLDQII PRMTTQSPLN QSSMNAMGSL SVLSPDRVLP QLISTITA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
Acts as a positive activator of the EIF2AK4/GCN2 protein kinase activity in response to amino acid starvation. Forms a complex with EIF2AK4/GCN2 on translating ribosomes; during this process, GCN1 seems to act as a chaperone to facilitate delivery of uncharged tRNAs that enter the A site of ribosomes to the tRNA-binding domain of EIF2AK4/GCN2, and hence stimulating EIF2AK4/GCN2 kinase activity. Participates in the repression of global protein synthesis and in gene-specific mRNA translation activation, such as the transcriptional activator ATF4, by promoting the EIF2AK4/GCN2-mediated phosphorylation of eukaryotic translation initiation factor 2 (eIF-2-alpha/EIF2S1) on 'Ser-52', and hence allowing ATF4-mediated reprogramming of amino acid biosynthetic gene expression to alleviate nutrient depletion. [UniProt].
Specifications
| GCN1L1 | |
| Polyclonal | |
| Unconjugated | |
| GCN1 | |
| 4932409G22; AL022764; eIF-2-alpha kinase activator GCN1; G431004K08Rik; Gcn1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF2 alpha kinase activator homolog; GCN1 eIF-2-alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1 general control of amino-acid synthesis 1-like 1 (yeast); GCN1, eIF2 alpha kinase activator homolog; GCN1L; GCN1L1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; hsGCN1; KIAA0219; mKIAA0219; peroxisome proliferator activated receptor interacting complex protein; PRIC295; translational activator GCN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92616 | |
| GCN1 | |
| Recombinant protein corresponding to Human GCN1L1. Recombinant protein control fragment (Product #RP-92376). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction