missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33040
This item is not returnable.
View return policy
Description
GCM2 Polyclonal specifically detects GCM2 in Human samples. It is validated for Western Blot.
Specifications
| GCM2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GCM2 | |
| Recombinant protein encompassing a sequence within the center region of human GCM2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9247 | |
| Human | |
| IgG |
| Western Blot | |
| 0.75mg/mL | |
| Western Blot 1:500-1:3000 | |
| chorion-specific transcription factor GCMb, GCM motif protein 2, GCMBglial cells missing homolog b, glial cells missing (Drosophila) homolog b, Glial cells missing homolog 2, glial cells missing homolog 2 (Drosophila), glide/gcm protein homolog, hGCMbglial cells missing 2 | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction