missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21724-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.Specifications
| GCM1 | |
| Polyclonal | |
| Unconjugated | |
| GCM1 | |
| GCM motif protein 1, GCM1, GCMA, Glial cells missing homolog 1, hGCMa | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8521 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NP62 | |
| GCM1 | |
| GCM1 Fusion Protein Ag16383 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction