missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCLM Recombinant Rabbit Monoclonal Antibody (23GB5535), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553894
This item is not returnable.
View return policy
Description
Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
Specifications
| GCLM | |
| Recombinant Monoclonal | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O09172, P48507, P48508 | |
| Gclm | |
| A synthesized peptide derived from human GCLM (65-100AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5535 | |
| Unconjugated | |
| Gclm | |
| AI649393; gamma glutamylcysteine synthetase; gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; Gclm; Gcmc; GCS light chain; Glclr; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; glutamate cysteine ligase, modifier subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate--cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase, modifier subunit; GSC light chain; null | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14630, 2730, 29739 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction