missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCLM Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
£119.00 - £330.00
Specifications
| Antigen | GCLM |
|---|---|
| Clone | 4K16 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235587
|
Proteintech
82623-1-RR-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233876
|
Proteintech
82623-1-RR-100UL |
100 μL |
£360.00 £330.00 / 100µL Save £30.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GCLM antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human GCLM. This antibody recognizes human, mouse, rat antigen. The GCLM antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| GCLM | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 2730 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 4K16 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| GLCLR | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts