missing translation for 'onlineSavingsMsg'
Learn More

GCLM Rabbit anti-Human, Clone: 0U3J2, Novus Biologicals™

Product Code. 18335214
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335214 100 μg 100µL
18303173 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18335214 Supplier Novus Biologicals Supplier No. NBP315380100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GCLM Monoclonal antibody specifically detects GCLM in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCLM
Applications Western Blot
Classification Monoclonal
Clone 0U3J2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Gamma-ECS regulatory subunit, Gamma-glutamylcysteine synthetase regulatory subunit, GCS light chain, GLCLRglutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory(30.8kD), Glutamate--cysteine ligase modifier subunit, glutamate-cysteine ligase regulatory protein, glutamate-cysteine ligase, modifier subunit, GSC light chain
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 175-274 of human GCLM (P48507).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2730
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.