missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCLM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35408-20ul
This item is not returnable.
View return policy
Description
GCLM Polyclonal antibody specifically detects GCLM in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| GCLM | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Gamma-ECS regulatory subunit, Gamma-glutamylcysteine synthetase regulatory subunit, GCS light chain, GLCLRglutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory(30.8kD), Glutamate--cysteine ligase modifier subunit, glutamate-cysteine ligase regulatory protein, glutamate-cysteine ligase, modifier subunit, GSC light chain | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-274 of human GCLM (NP_002052.1).,, Sequence:, MGTDSRAAKALLARARTLHLQTGNLLNWGRLRKKCPSTHSEELHDCIQKTLNEWSSQINPDLVREFPDVLECTVSHAVEKINPDEREEMKVSAKLFIVESNSSSSTRSAVDMACSVLGVAQLDSVIIASPPIEDGVNLSLEHLQPYWEELENLVQSKKIVAIGTSDLDKTQLEQLYQWAQVKPNSNQVNLASCCVMPPDLTAFAKQFDIQLLTHNDPKELLSEASFQEALQESIPDIQAHEWVPLWLLRYSVIVKSRGIIKSKGYILQAKRRGS | |
| 20 μL | |
| Cancer | |
| 2730 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction