Learn More
Invitrogen™ GCLC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587854
Description
Immunogen sequence: MGLLSQGSPL SWEETKRHAD HVRRHGILQF LHIYHAVKDR HKDVLKWGDE VEYMLVSFDH ENKKVRLVLS GEKVLETLQE KGERTNPNHP TLWRPEYGSY MIEGTPGQPY GGTMSEFNTV EANMRKRRKE ATSILEENQA LCTITSFPRL GCPGFTLPEV KPNPVEGGAS KSLFFPDEAI NKHPRFSTLT RNIRHRRGEK VVINVPIFKD KNTPSPFIET FTEDDEASRA SKPDHIYMDA MGFGMGNCCL QV; Positive Samples: Mouse kidney, Mouse liver, Mouse lung, Rat liver.
Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
Specifications
| GCLC | |
| Polyclonal | |
| Unconjugated | |
| Gclc | |
| cb1049; D9Wsu168e; EGK_15023; fe36e11; gamma GCS-HS; gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCL; Gclc; GCS; GCS heavy chain; Ggcs-hs; GLCL; GLCLC; GLCL-H; glutamate-cysteine ligase catalytic subunit; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain; wu:fe36e11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14629, 25283, 2729 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.66 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P19468, P48506, P97494 | |
| Gclc | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-252 of human GCLC (NP_0014891). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.