Learn More
GCLC Antibody (S08-2F5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19644-100ul
 View more versions of this product
Product Code. 18646853
  • £500.00 / 100µL
Description
GCLC Monoclonal antibody specifically detects GCLC in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GCLC
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S08-2F5
Unconjugated
EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit
A synthetic peptide of human GCLC (Uniprot # P48506)
100 μg
Cell Biology, Lipid and Metabolism
2729
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.