missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCKR Antibody (OTI1E7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70823F
This item is not returnable.
View return policy
Description
GCKR Monoclonal specifically detects GCKR in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GCKR | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| GCKR | |
| Full length recombinant protein of human GCKR (NP_001417) produced in HEK293T cells. | |
| 0.1 mL | |
| 2646.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E7 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FGQTL5, GKRP, glucokinase (hexokinase 4) regulator, glucokinase (hexokinase 4) regulatory protein, Glucokinase regulator, glucokinase regulatory protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction